Learn More
Bochem™ Stainless Steel Conical Tube Funnel
Product Code. 10788931
Change view
Click to view available options
Diameter (Metric) Stem:
120 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Stem Unit Size
10788931 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10788931 Supplier Bochem™ Supplier No. 8842
Estimated Shipment: 03-12-2026
Manufactured from 18/10 stainless steel. BOCHEM™ Stainless Steel Conical Tube Funnel will help keep your work areas clean and avoid waste.
  • With air canal
  • Handle 
  • Tube conical shape 
Specifications
Diameter (Metric) Stem 120 mm
Material Stainless Steel
Height (Metric) 145 mm