Learn More
Cole-Parmer™ Stainless Steel Compression Fittings
Product Code. 11835453
Change view
Click to view available options
For Use With:
0.125 in. NPT(M)
0.25 in. NPT(M)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11835453 0.125 in. NPT(M) Each
11845453 0.25 in. NPT(M) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11835453 Supplier Cole-Parmer™ Supplier No. 3140634
  • ONLINE EXCLUSIVE PRICE
    20.15 EUR
    15.75 EUR / Pack of 1
    Save 4.40 EUR
    21% Off
Estimated Shipment: 21-08-2026
Specifications
Autoclavable Autoclavable
Material Stainless Steel
For Use With 0.125 in. NPT(M)
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 1 Pack
Max. Operating Temperature (Metric) 93°C
Product Type Compression Fitting