Learn More
SI Analytics™ Stainless Steel Brackets
Product Code. 10751201 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
Reverse Flow Viscometer
Ubbelohde Viscometer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10751201 Ubbelohde Viscometer Each
10241881 Reverse Flow Viscometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10751201 Supplier SI Analytics™ Supplier No. 285405043
Only null left
Add to basket
Add to basket
Protect your viscometer from breakage. SI Analytics™ Stainless Steel Brackets are designed to ensure that the viscometers are held in a vertical position.
  • The maximum deviation is < 1°
  • In application in conjunction with SCHOTT Instruments and other commercially available see-through thermostats the viscometers can only be used with the appropriate stand or bracket
  • Provide protection for viscometers
  • Allow viscometers to be held vertically
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bracket
For Use With (Equipment) Ubbelohde Viscometer
Material Stainless Steel
For Use With (Application) For manual and automatic measurements