Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SN75, 15 mm
Product Code. 17568745 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN75 Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17568745 SN75 Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17568745 Supplier Memmert™ Supplier No. SN75B04358
Estimated Shipment: 17-11-2026
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN75 Sterilizer
Product Type Bottom Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel