Learn More
Bochem™ Stainless Steel Alcohol Burner
Product Code. 10577191
Change view
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10577191 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10577191 Supplier Bochem™ Supplier No. 7200
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 04-12-2026
Specifications
Product Type Alcohol Burner
Material Stainless Steel
Capacity (Metric) 150 mL
Diameter (Metric) 85 mm
Height (Metric) 65 mm
No. per Pack 1 Ea.
Weight (Metric) 0.1 kg