Learn More
Thermo Scientific™ Stained Salmonella O and H Suspensions
Product Code. p-8401005
Change view
Click to view available options
Detectable Analytes:
S. paratyphi B-O
S. paratyphi C-H phase 1
S. typhi-H
Unit Size:
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
13235729 S. typhi-H 5mL
13255729 S. paratyphi C-H phase 1 5mL
11913272 S. paratyphi B-O 5mL
13275729 S. typhi-H 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13235729 Supplier Thermo Scientific™ Supplier No. R30855001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rapidly detect and quantify specific antibodies to Salmonella in sera with this standardized stained suspension.
Rapidly detect and quantify specific antibodies to Salmonella spp. in sera with Thermo Scientific™ Remel™ Stained Salmonella O and H Suspensions. Stained Salmonella O and H Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.
Use Stained Salmonella O and H Suspensions for the qualitative detection of Salmonella antibodies in sera for epidemiological and diagnostic purposes, primarily in the investigation of enteric infections and pyrexia.
  • Identifies major groups of Salmonella serotypes
  • Provides accurate, reliable results
  • Simple, easy-to-use tests
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens. In the diagnosis of febrile disease such tests should be used in parallel with appropriate cultural techniques for the isolation and identification of the causative organism. The suspensions are suitable for use in rapid slide tests1 and standard Widal tests2, 3. They may also be used as control antigens for serological identification of bacterial isolates.
 Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Stained Salmonella O and H Suspensions
Detectable Analytes S. typhi-H
Type Salmonella O and H Suspensions
Includes Stained Salmonella O and H Suspensions, Instruction for Use
Quantity 5 mL
Target Organism Class Salmonella