Learn More
Eppendorf™ Stacking and Leveling
Stacking and leveling Incubators/Shakers
70.90 EUR - 128.00 EUR
Specifications
For Use With (Equipment) Incubators and Shakers
View More Specs
Products 3
Product Code Brand For Use With (Application) Price Quantity & Availability  
Product Code Brand For Use With (Application) Price Quantity & Availability  
15713695
View Documents
Eppendorf™
2277000006
Stack and Level <100 kg
70.90 EUR
Pack of 1
View alternative products
Save Up to 
»
17161066
View Documents
Eppendorf™
2277000012
Stack and Level 100 to 180 kg
99.25 EUR
Pack of 1
View alternative products
Save Up to 
»
17171066
View Documents
Eppendorf™
2277000013
Stack and Level >180 kg
128.00 EUR
Pack of 1
View alternative products
Save Up to 
»
Description
Description
  • Stacking and leveling Incubators/Shakers
Specifications
Specifications
Incubators and Shakers