Learn More
Kartell SpA Stackable Deep Tray
Product Code. 12071103
Change view
Click to view available options
Capacity (Metric):
10 L
16 L
20 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12071103 10 L Each
11381105 16 L Each
12081103 20 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12071103 Supplier Kartell SpA Supplier No. 0054200
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 01-12-2026
Deep tray, low form, stackable
Stackable deep trays made of HDPE with handles
Specifications
Capacity (Metric) 10 L
Closure Type None
Color White
Material HDPE
Shape Rectangular
Product Type Deep Tray
Sterility Non-sterile
Dimensions (L x W x H) 430 x 310 x 100 mm
Length (Metric) 430 mm
Width (Metric) 310 mm
Height (Metric) 100 mm