Learn More
KD Scientific™ SS Syringe, 1/16 in Swagelok Fitting
Product Code. 18811675
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
18811675 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18811675 Supplier KD Scientific™ Supplier No. 780806
  • ONLINE EXCLUSIVE PRICE
    2105.30 EUR
    1920.71 EUR / Pack of 1
    Save 184.59 EUR
    8% Off
Estimated Shipment: 14-01-2027
Durable, electron-beam construction withstands high force, high pressure applications
  • Eliminates hazards of glass syringe breakage
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge No Needle Included
Latex Free Yes
Disposable Reusable
Volume (Metric) 200 mL
For Use With (Equipment) KD Scientific Syringe Pumps
Connection Type Swagelok
Graduated No