missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SRT 1720
GREENER_CHOICE

Product Code. 18707896
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18707896

Brand: Tocris Bioscience™ 7558/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective SIRT1 activator; induces autophagy and mitochondrial biogenesis

SRT 1720 is selective SIRT 1 activator (EC50 = 0.16 μM). SRT 1720 inhibits growth of bladder cancer organoids and xenografts (IC50 = 0.35 μM) and induces autophagy. In vitro, SRT 1720 increases mitochondrial biogenesis and recovery after oxidation injury and reduces palmitate-induced insulin resistance. Also improves survival and healthspan of obese mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[3-(1-Piperazinylmethyl)imidazo[2,1-b]thiazol-6-yl]phenyl]-2-quinoxalinecarboxamide
CAS 925434-55-5
Quantity 10 mg
Target Sirtuin 1/SIRT1
Molecular Formula C25H23N7OS
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.