Learn More
Bochem™ Square Wire Basket
Description
- Square
- Special high quality processing
- 18/10 E-Poli
Specifications
Specifications
| Material | 18/10 E-Poli |
| Shape | Square |
| Size | 8 x 8 x 1 mm Mesh |
| Dimensions (L x W x H) | 100 x 100 x 100 mm |
Product Title
Haben Sie Verbesserungsvorschläge?