missing translation for 'onlineSavingsMsg'
Learn More

SPZ1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

199.00 EUR - 351.00 EUR

Specifications

Antigen SPZ1
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18140582
View Documents
Novus Biologicals
NBP2-24582SS
0.025 mg
199.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18717283
View Documents
Novus Biologicals
NBP2-24582
0.1 mg
351.00 EUR
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SPZ1 Polyclonal specifically detects SPZ1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SPZ1
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Rabbit
Human, Mouse, Rat, Primate
Q9BXG8
84654
A portion of amino acids 40-90 of human SPZ1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
FLJ25709, NYD-TSP1, spermatogenic leucine zipper 1, spermatogenic leucine zipper protein 1, spermatogenic zip 1, Testis-specific protein 1, Testis-specific protein NYD-TSP1, TSP1
SPZ1
IgG
Affinity Purified
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in rat, mouse, monkey and dog, and 82% homologous in chicken.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.