Learn More
SPZ1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-24582
 View more versions of this product
Product Code. 18717283
  • 351.00 EUR / 0.10mg
Description
SPZ1 Polyclonal specifically detects SPZ1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
A portion of amino acids 40-90 of human SPZ1 was used as the immunogen.
0.1 mg
Rat, Primate
IgG
Rabbit
Affinity Purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Certificates
Promotions
Promotions
For Research Use Only