Learn More
SPZ1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24582
This item is not returnable.
View return policy
Description
SPZ1 Polyclonal specifically detects SPZ1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| A portion of amino acids 40-90 of human SPZ1 was used as the immunogen. | |
| 0.1 mg | |
| Rat, Primate | |
| IgG |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |