Learn More
Edulab Spring Extension, Steel, Pack of 200
Product Code. 30280265
Change view
Click to view available options
Product Type:
Replacement Spring
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30280265 Replacement Spring Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280265 Supplier Edulab Supplier No. VTN12302388
  • ONLINE EXCLUSIVE PRICE
    69.30 EUR
    67.21 EUR / Pack of 200
    Save 2.09 EUR
    3% Off
Estimated Shipment: 09-12-2026
Coiled steel spring with double loop ends, ideal for physics experiments and motion studies.
  • Steel coiled spring with double loop ends for secure attachment
  • End-to-end length of 50 mm (1.96 in.) and collapsed length of 24 mm (0.94 in.) without hoops
  • Outside diameter of 15 mm (0.59 in.)
  • Extends approximately 200-230 mm (8-9 in.) and returns to original length without deformity
  • Ideal for studying motion of mass on a spring, Hooke’s Law, potential energy, and related physics concepts
  • Provides a visually and kinesthetically effective method for learning spring dynamics
  • Single spring included for individual experiments
Specifications
Product Type Replacement Spring