Learn More
Tocris Bioscience™ SPP 86
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SPP 86 is a potent RET inhibitor (IC50 = 8 nM). Also exhibits inhibitory activity at EphA1, FGFR1, Flt4, Lck and Yes. Inhibits proliferation of HCT 116 and TPC1 cells in vitro. Cell permeable.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | SPP 86 |
| Purity | >98% |
| Molecular Weight (g/mol) | 277.32 |
For Research Use Only