Learn More
Spiroxamine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Spiroxamine-d4 |
| CAS | 2469272-82-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H31D4NO2 |
| Quantity | 5 mg |
| Synonym | 8-(1,1-dimethylethyl)-N-ethyl-N-propyl-1,4-dioxaspiro[4.5]decane-2-methanamine-6,6,10,10-d4 |
| InChI Formula | InChI=1S/C18H35NO2/c1-6-12-19(7-2)13-16-14-20-18(21-16)10-8-15(9-11-18)17(3,4)5/h15-16H,6-14H2,1-5H3/i10D2,11D2 |
| SMILES | CCN(CC1COC2(C([2H])([2H])CC(C(C)(C)C)CC2([2H])[2H])O1)CCC |
| IUPAC Name | N-((8-(tert-butyl)-1,4-dioxaspiro[4.5]decan-2-yl-6,6,10,10-d4)methyl)-N-ethylpropan-1-amine |
| Molecular Weight (g/mol) | 301.5 |
| Show More |