Learn More
Spirotetramat-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Spirotetramat ethyl-d5 |
| CAS | 2469007-09-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21H22D5NO5 |
| Quantity | 5 mg |
| Synonym | carbonic acid','cis-3-(2,5-dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl ethyl ester-d5 |
| InChI Formula | InChI=1S/C21H27NO5/c1-5-26-20(24)27-18-17(16-12-13(2)6-7-14(16)3)19(23)22-21(18)10-8-15(25-4)9-11-21/h6-7,12,15H,5,8-11H2,1-4H3,(H,22,23)/t15-,21+/i1D3,5D2 |
| SMILES | O=C1C(C2=C(C)C=CC(C)=C2)=C(OC(OC([2H])([2H])C([2H])([2H])[2H])=O)[C@]3(CC[C@H](OC)CC3)N1 |
| IUPAC Name | (5s,8s)-3-(2,5-dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl (ethyl-d5) carbonate |
| Molecular Weight (g/mol) | 378.48 |
| Show More |