missing translation for 'onlineSavingsMsg'
Learn More

Spinosin, TRC

Product Code. 30136443
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136443 100 mg 100mg
30011798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136443 Supplier Toronto Research Chemicals Supplier No. TRCS681950100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72063-39-9
Molecular Formula C28 H32 O15
Molecular Weight (g/mol) 608.54
Synonym 4H-1-Benzopyran-4-one, 6-(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-, 6-(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one, Flavoayamenin, Spinosin, Swertisin 2' '-O-glucoside
IUPAC Name 6-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxychromen-4-one
SMILES COc1cc2OC(=CC(=O)c2c(O)c1[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c5ccc(O)cc5

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C28H32O15/c1-39-14-7-15-18(12(32)6-13(40-15)10-2-4-11(31)5-3-10)22(35)19(14)26-27(24(37)21(34)16(8-29)41-26)43-28-25(38)23(36)20(33)17(9-30)42-28/h2-7,16-17,20-21,23-31,33-38H,8-9H2,1H3/t16-,17-,20-,21-,23+,24+,25-,26+,27-,28+/m1/s1
Formula Weight 608.1741
Chemical Name or Material Spinosin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.