CAS RN 72063-39-9
CAS RN 72063-39-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Spinosin, TRC
CAS: 72063-39-9 Molecular Formula: C28 H32 O15 Molecular Weight (g/mol): 608.54 Synonym: 4H-1-Benzopyran-4-one, 6-(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-,6-(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one,Flavoayamenin,Spinosin,Swertisin 2' '-O-glucoside IUPAC Name: 6-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]-5-hydroxy-2-(4-hydroxyphenyl)-7-methoxychromen-4-one SMILES: COc1cc2OC(=CC(=O)c2c(O)c1[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c5ccc(O)cc5