Learn More
Heidolph™ Spider with Flasks
Product Code. 10688382 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10688382 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10688382 Supplier Heidolph™ Supplier No. 1530000504
Estimated Shipment: 28-12-2026
For use with rotary evaporators
Comes with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Type Glassware Assembly Rotary Evaporator
Volume (Metric) 50 mL
Voltage 115/230 VAC