Description
Ideal for samples which crumble or ablate after pressing.
- Select binders from powder, tablet, or liquid form depending on your application.
- High-quality sample preparation is the key to achieving consistent and reliable analytical results.
- Powders are ideal for samples that crumble or ablate after pressing.
- They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
- Tablets are an efficient way to add binder to XRF sample disks.
- Liquid easily mixes with the sample.
Specifications
Specifications
| For Use With (Equipment) | SamplePrep Mixers, SamplePrep Mills |
| Type | Cellulose Binder |
| For Use With (Application) | Samples Which Crumble or Ablate After Pressing |
| Particle Size | ≤20 μm |
| Quantity | 150 g |