Learn More
Cole-Parmer™ XRF Spec-Caps™ Cups
Product Code. 15300552 Shop All SPEX Certiprep Products
Change view
Click to view available options
Diameter (Metric):
30 mm
31 mm
33 mm
38 mm
Height (Metric):
8 mm
9 mm
Quantity:
100 Pk.
1000 Pk.
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) Quantity unitSize
15300552 38 mm 9 mm 100 Pk. Pack of 100
15360542 33 mm 8 mm 100 Pk. Pack of 100
15310552 38 mm 9 mm 1000 Pk. Pack of 1000
15390542 31 mm 8 mm 1000 Pk. Pack of 1000
15380542 31 mm 8 mm 100 Pk. Pack of 100
15340542 30 mm 8 mm 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15300552 Supplier Cole-Parmer™ Supplier No. 3617
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
  • Reinforces sample disks to resist chipping and breaking.
  • Makes sample disks easier to remove from the pellet die.
  • Minimizes contamination during sample handling.
  • Protects the bore of the pellet die from abrasive samples.
  • Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Specifications
For Use With (Equipment) XRF Spectrometers
Type Sample Cup
Description Produces pellets 40 mm x 5 mm
Diameter (English) 1.5 in.
Diameter (Metric) 38 mm
Height (English) 0.35 in.
Height (Metric) 9 mm
Material Aluminum
Quantity 100 Pk.