Learn More
SPEX CertiPrep™ OdorEroder
Product Code. 15296106 Shop All SPEX Certiprep Products
Change view
Click to view available options
Volume (Metric):
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15296106 250 g 250g
15286106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15296106 Supplier SPEX CertiPrep™ Supplier No. ODER250G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. Afficher la politique du retour.
Règlementaire: Ce produit contient un précurseur de drogue de catégorie 2 selon le règlement (CE) n ° 273/2004, tout client doit remplir une déclaration avant la livraison.
Effectively neutralizes a wide range of chemical odors in the lab
  • Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
  • As odors get trapped, the beads start turning brown
Spécification
For Use With (Application) Effectively neutralizes a wide range of chemical odors in the lab
Volume (Metric) 250 g