Learn More
RETSCH Spektromelt™ C20 Cellulose Tablet
Brand: RETSCH 22.440.0003
39083.00 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.10000kg
Specifications
Spektromelt C20 Cellulose Tablet | |
P 25 and PP 40 Pellet Presses | |
1 kg |