missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Speedglas™ Welding Helmet G5-03 Pro with Welding Filter G5-01/03TW
Description
- Designed for versatile welders who need seamless switching between welding and grinding tasks
- Tap functionality allows quick activation of grind or cut mode with a simple triple tap anywhere on the helmet
- Offers highly customisable adjustment settings to tailor visibility and comfort to the user
- Provides enhanced control over viewing the weld pool and configuring protection against radiation, sparks and spatter
- Features 3M™ Speedglas™ Natural Color Technology for improved clarity
- Includes Variable Color Technology with G5-01/03VC filters and Tack Welding Mode with G5-01/03TW (631820)
- Compatible with an optional Task Light to improve visibility of the workpiece
- Supports an optional helmet adapter system for connecting a hard hat when extra head protection is required
- Built with next-generation technology to deliver dynamic, responsive welding protection
- Intuitive design supports the demands of challenging welding environments
- Can be equipped with various accessories, such as a task light, neck cover and head cover, for personalised protection
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Welder Adapter |
| Product Type | Welding Helmet |
| Size | Universal |
| Ventilated | Not Ventilated |
| Head Style | Ratchet |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?