Learn More
3M™ Speedglas™ Visor Plate (Standard)
Product Code. 15839634 Shop All 3M Products
Change view
Click to view available options
Lens Tint: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15839634 Lens Tint: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15839634 Supplier 3M™ Supplier No. 7000000236
  • ONLINE EXCLUSIVE PRICE
    86.00 EUR
    66.66 EUR / Pack of 5
    Save 19.34 EUR
    22% Off
Estimated Shipment: 24-02-2027
3M™ Speedglas™ Visor Plate (Standard) is a replacement part for use with Speedglas™ Welding Helmets 9100 FX, 9100 FX-Air, and 9100 MP.
  • Replacement spare part to the original visor plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
  • Regularly clean with a soft cloth
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Speedglas Welding Masks 9100 FX, 9100 FX-Air and 9100 MP
Lens Tint Clear
Product Type Lens
Quantity 5/Pk.