Learn More
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide
Product Code. 11528692 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11528692 5 mL Each
11854143 100 mL Each
11864143 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11528692 Supplier Cytiva Supplier No. 78152104010150
  • ONLINE EXCLUSIVE PRICE
    996.00 EUR
    794.21 EUR / Pack of 1
    Save 201.79 EUR
    20% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Capture of Biotinylated Probes. Their cauliflower-like structure gives Neutravidin beads high binding capacity. Neutravidin SpeedBeads combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in immunoassay and molecular biology applications.
  • Neutravidin beads exhibit high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond quickly to a magnetic field to shorten assay times, improve precision and move faster through viscous solutions
  • Available with a 3500-4500 pmol/mg biotin-binding range
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Includes 0.05% Na azide
Quantity 5 mL
Diameter (Metric) 1 μm