Description
- PC/TPR Co-injected frame for optimum comfort.
- Multiple coatings and sizes available.
- Soft gasket to fit most face structures.
- Optimal panoramic vision.
Specifications
Specifications
| Lens Coating | Platinum |
| Lens Tint | Smoke |
| Product Type | Safety Goggles |
| Model | SPECTN20W |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| UV Protection | None |
| Frame Type | Indirect Vented |
| Show More |