Learn More
Spectromelt™ A 1000, Supelco™
Product Code. 15845660
Change view
Click to view available options
Includes:
Powder, 1 kg
Powder, 5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15845660 Powder, 5 kg 5kg
15835660 Powder, 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15845660 Supplier Supelco Supplier No. 1131755000
 Restricted Item
Estimated Shipment: 15-01-2027
For X-ray fluorescence analysis, Li2B4O7 + 0.07% LiBr
TRUSTED_SUSTAINABILITY
Chemical Identifiers
For Use With (Application) X-ray Fluorescence Analysis
Specifications
Includes Powder, 5 kg