missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta III (SPTBN2) Mouse, Clone: SPTBN2/1778, enQuireBio™
Specifications
Specifications
| Antigen | Spectrin beta III (SPTBN2) |
| Applications | Immunohistochemistry, ELISA |
| Cellular Localization | Cell Surface |
| Classification | Monoclonal |
| Clone | SPTBN2/1778 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. |
| Host Species | Mouse |
| Immunogen | Recombinant fragment of human Spectrin beta III (SPTBN2) protein (apprx. aa 356-475) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?