Learn More
Molecular Devices SpectraDrop™ Micro‐Volume HTS Kit for SpectraMax™ Microplate Readers
Shop All Molecular Devices ProductsDescription
- Enables high-throughput micro-volume nucleic acid and protein quantification workflows
- Supports low-volume measurements for UV-Vis spectrophotometry applications
- Measures sample volumes as low as 2 μL
- Includes one microplate adapter, multiple 24-well and 64-well low-volume bottom slides, and 2 μL and 4 μL cover slides
- Supports both 24-well and 64-well configurations for increased assay throughput
- Uses a slide-based design with pathlength control for consistent and reproducible analysis
- Supports microplate-based assay workflows with flexible density configurations
- Designed for use with compatible SpectraMax microplate readers
Specifications
Specifications
| Product Type | Accessory Kit |
| For Use With (Equipment) | Microplate Reader |
| Sterility | Non-sterile |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?