missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Dihydrochloride, TRC

Product Code. 30133709
Change view
Click to view available options
Quantity:
100 mg
500 mg
10 g
Unit Size:
100mg
10g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133709 10 g 10g
17962085 100 mg 100mg
17933665 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133709 Supplier Toronto Research Chemicals Supplier No. TRCS68025010G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21736-83-4
Molecular Formula C14 H24 N2 O7 . 2 Cl H
Molecular Weight (g/mol) 405.2714
Synonym 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, hydrochloride (1:2), (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-, 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, dihydrochloride, (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)- (9CI), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one, decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-, dihydrochloride, [2R-(2α,4aβ,5aβ,6β,7β,8β,9α,9aα,10aβ)]- (8CI), (+)-Spectinomycin dihydrochloride, NSC 248616, Spectinomycin dihydrochloride, Spectinomycin hydrochloride, Stanilo
SMILES Cl.Cl.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H24N2O7.2ClH/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;/m1../s1
Formula Weight 404.1117
Chemical Name or Material Spectinomycin dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.