missing translation for 'onlineSavingsMsg'
Learn More

Melford Spectinomycin Dihydrochloride Pentahydrate

Product Code. p-200087009
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018773 1 g 1g
30018775 5 g 5g
30018774 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018773 Supplier Melford Supplier No. S230001.0
Add to basket
Add to basket
This item is not returnable. View return policy

Spectinomycin dihydrochloride pentahydrate is an antibiotic that is commonly used in the life science marketplace.

Spectinomycin dihydrochloride pentahydrate is an antibiotic that is commonly used in the life science marketplace. It belongs to the class of aminocyclitol antibiotics and is primarily utilized for its activity against gram-negative bacteria.

Spectinomycin dihydrochloride pentahydrate finds applications in various fields such as research, molecular biology, and biotechnology. It is commonly used as a selective agent in cell culture systems and is added to growth media to prevent the growth of bacteria that may contaminate cell lines.

Also, it is particularly effective against certain strains of gram-negative bacteria, including Escherichia coli. It works by inhibiting protein synthesis in bacteria, which ultimately leads to their death. Its selective action makes it valuable for maintaining sterile conditions and selecting cells that have taken up foreign genetic material in molecular biology experiments.

Specifications

Chemical Name or Material Spectinomycin Dihydrochloride Pentahydrate
CAS 22189-32-8
Quantity 1 g
Molecular Formula C14H24N2O7-2 HCl-5H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.