Learn More
asecos™ Special Sorbents Mat
Product Code. 17816396 Shop All asecos Products
Change view
Click to view available options
Weight (Metric):
6.4 kg
7.3 kg
Förpackningsstorlek:
Pack of 100
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Weight (Metric) unitSize
17816396 6.4 kg Pack of 100
17846396 7.3 kg Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17816396 Leverantör asecos™ Leverantörsnummer 9917
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Denna artikel kan inte returneras. Se returpolicy
The Special Sorbents Mat is ideal for the safe absorption of acids, alkalis, aggressive chemicals and oil.
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibers, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifikationer
Color Yellow
Material Heavy-duty Cloth
Type Chemical Sorbent Pad
Length (English) 15.75 in.
Length (Metric) 400.00 mm
Width (English) 15.75 in.
Width (Metric) 400.00 mm
Weight (English) 14.11 lb.
Weight (Metric) 6.4 kg
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.