Learn More
Brady™ SPC™ Absorbents SitePoint Maintenance Kit
Product Code. 17861755 Shop All Brady Products
Change view
Click to view available options
Capacity (English):
26.4 gal.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Unit Size
17861755 26.4 gal. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17861755 Supplier Brady™ Supplier No. 309079
  • ONLINE EXCLUSIVE PRICE
    1085.80 EUR
    915.93 EUR / 1 sets
    Save 169.87 EUR
    15% Off
Estimated Shipment: 09-12-2026
Ideal for moving spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required, which is especially ideal for maintenance activities, at construction sites and other temporary locations.
  • Loaded SitePoint Mobile Unit with suitable products for cleaning up general spills
  • Features two (200 mm) wheels for mobility and anchor points for fixed installation
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
  • Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission
Specifications
Color Yellow, White
Capacity (English) 26.4 gal.
Capacity (Metric) 100 L
Material MDPE
Type Spill Kit
Format Spill Kit
For Use With (Application) Beverage and Food Processing, Chemical, Healthcare, Maritime, Oil and Gas, Pharmaceutical, Warehouse
Includes SitePoint Mobile Unit, ALLWICK Universal Refill, Wall-mounted Roll Dispenser (50 cm Wide), Storage Unit for Gloves, Storage Unit for Goggles, A4 Wall Document Holder, Fixings
Length (English) 2.3 in.
Length (Metric) 5.95 cm
Width (English) 29.724 in.
Width (Metric) 75.5 cm
Height (Metric) 172.5 cm
Depth (Metric) 5.95 cm
Weight (English) 35.3 lb.
Weight (Metric) 16 kg
Alternate Part Number SPHS1-BB1-YE-MAINT