Learn More
Brady™ SPC™ Absorbents SitePoint Chemical Maintenance Kit
Click to view available options
Capacity (English):
26.4 gal.
Conditionnement:
1 set
Description
- Loaded SitePoint Mobile Unit with suitable products for cleaning up general spills
- Features two (200 mm) wheels for mobility and anchor points for fixed installation
- Fix items to backboard using self-tapping screws or bolts
- Suitable for inside or outdoor use
- Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission
Spécification
Spécification
| Color | Yellow, White |
| Capacity (English) | 26.4 gal. |
| Capacity (Metric) | 100 L |
| Material | MDPE |
| Type | Spill Kit |
| Format | Spill Kit |
| For Use With (Application) | Beverage and Food Processing, Chemical, Healthcare, Maritime, Oil and Gas, Pharmaceutical, Warehouse |
| Includes | SitePoint Mobile Unit, Chemical Refill, Wall-mounted Roll Dispenser (50 cm Wide), Storage Unit for Gloves, Storage Unit for Goggles, A4 Sign Holder, Fixings |
| Length (English) | 2.3 in. |
| Length (Metric) | 5.95 cm |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu