Learn More
BRAND™ Spare Burette Tip
Product Code. 10626483 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10626483 25 mL Each
10434352 10 mL Each
10502423 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10626483 Supplier BRAND™ Supplier No. 11511
  • ONLINE EXCLUSIVE PRICE
    7.00 EUR
    6.30 EUR / Pack of 1
    Save 0.70 EUR
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-12-2026
For compact and automatic burettes
  • Clear Boro 3.3 glass
  • For use with compact burettes and compact automatic burettes
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity (Metric) 25 mL
    Material Borosilicate Glass (3.3)
    No. per Pack 1
    Volume (Metric) Reservoir 25 mL