missing translation for 'onlineSavingsMsg'
Learn More

SPADNS

Product Code. p-7023410
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11428766

Brand: Thermo Scientific Alfa Aesar A16933.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23647-14-5 | C16H10N2Na3O11S3+ | 571.408 g/mol

Sulfanilic acid azochromotrop is used as an indicator for complexometric determinations of zirconium and thorium. It is used as a reagent for drinking and wastewater analysis using Hach Method 8029, Fluoride, SPADNS Method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfanilic acid azochromotrop is used as an indicator for complexometric determinations of zirconium and thorium. It is used as a reagent for drinking and wastewater analysis using Hach Method 8029, Fluoride, SPADNS Method.

Solubility
Soluble in water (10 mg/ml), and ethanol (1 mg/ml). very slightly soluble or insoluble in alcohol and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23647-14-5
Molecular Formula C16H10N2Na3O11S3+
Molecular Weight (g/mol) 571.408
MDL Number MFCD00003952
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
Synonym Sulfanilic acid azochromotrope; 2-(4-Sulfophenylazo)chromotropic acid trisodium salt
PubChem CID 73658865
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300°C
Solubility Information Soluble in water (10mg/ml),and ethanol (1mg/ml). very slightly soluble or insoluble in alcohol and organic solvents.
Formula Weight 570.41
Quantity 10 g
Chemical Name or Material SPADNS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.