Learn More
Bel-Art™ Colony Counter Replacement Pen Tips
Product Code. 10414742
Change view
Click to view available options
Color:
Black
Red
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10414742 Black Each
12366417 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10414742 Supplier Bel-Art™ Supplier No. H378620100
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
Replacement pen tips fit Scienceware Colony Counter Cat. No. 07910018.
  • The long life, black marker tips work on plastic or glass and are oil and water resistant
Specifications
Product Type Replacement Pen Tip
Display Type LCD
Color Black
Length (English) 1.8 in.
Length (Metric) 4.6 cm
Width (English) 0.75 in.
Width (Metric) 1.9cm
For Use With (Application) Accurate sample counting, perfect for Plastic and Glass surfaces
For Use With (Equipment) Scienceware™ Electronic Colony Counter, F378620002