Learn More
Lenz Laborglasintrumente™ Soxhlet Extractors with Allihn Condenser
Product Code. 10346121
Change view
Click to view available options
Material:
DURAN Tubing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10346121 DURAN Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10346121 Supplier Lenz Laborglasintrumente™ Supplier No. 05365049
  • ONLINE EXCLUSIVE PRICE
    312.00 EUR
    256.77 EUR / Pack of 1
    Save 55.23 EUR
    17% Off
Estimated Shipment: 17-12-2026
For the extraction from solids with solvents
  • The solvent is recirculated continuously and distilled from the extracted sample before
    contacting the solid again
  • The extract is concentrated in the distillation flask
  • The method allows an effective separation with relatively small amounts of extracting solvents
  • The complete apparatuses (all made of DURAN™ tubing) are consisting of:
    • Extractor in compact and robust design, with flask connection NS 29/32
    • Allihn condenser with thread GL 14
    • Round bottom flask
Specifications
Material DURAN Tubing
Includes 250 mL extractor, Allihn condenser (NS 45/40), and 500 mL round bottom flask