Learn More
SOX17 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1924-SP
This item is not returnable.
View return policy
Description
SOX17 Polyclonal specifically detects SOX17 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.Product Title
Spot an opportunity for improvement?
For Research Use Only