Learn More
SOX17 Goat anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF1924-SP
Encompass
Product Code. 15791164
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
SOX17 Polyclonal specifically detects SOX17 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions