Learn More
3M™ Sound Examiner 400 Series Sound Level Meter SE 401
Description
- Sound Level Meter engineered to accurately measure noise levels in highly variable environments
- Intuitive interface allows for out of the box use with little or no training
- Preamp can be easily removed to enable interference-free low level monitoring
- Class 1
- Rechargeable lithium battery
- Store numerous noise surveys in memory for download
- Multilingual user interface
Specifications
Specifications
| Battery Life | 18 hours |
| Dimensions | 28.4 x 7.1 x 3.3 cm |
| Product Type | Sound Level Meter |
| For Use With (Application) | Area Monitoring (Sound Level Meter) |