Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Product Code. 10458191 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1 x 250 mL
1 x 750 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10458191 1 x 750 mL Each
10638952 1 x 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10458191 Supplier Thermo Scientific™ Supplier No. 75006441
  • ONLINE EXCLUSIVE PRICE
    487.00 EUR
    470.00 EUR / Pack of 1
    Save 17.00 EUR
    3% Off
In Stock
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor
Capacity 1 x 750 mL
Product Type Bucket
Description Round Bucket