Learn More
Sonoco™ ThermoSafe Envoy™ Global Pre-Qualified Insulated Shippers with Universal Packout
Product Code. 17848703
Change view
Click to view available options
Capacity (Metric):
2.5 L
12.1 L
Quantity:
24 Pallets
8 Pallets
Weight (Metric):
10.8 kg
22.3 kg
Unit Size:
Pack of 12
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Quantity Weight (Metric) unitSize
17848703 2.5 L 24 Pallets 10.8 kg Pack of 24
17838703 12.1 L 8 Pallets 22.3 kg Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17848703 Supplier Sonoco™ Supplier No. E3F4
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Temperature controlled packaging solution. Frozen (<-20°C).
  • Universal packout configuration
  • Positioned as low material packaging cost solution
  • Offers various sizes, temperature ranges and flexibility
  • Envoy Global line is able to change from controlled room temperature, refrigerated and frozen temperatures by changing the refrigerants
  • Simple and error-free
  • Utilizes four EPS container sizes
  • Pre-Qualified to ISCsilver ambient profile
  • Zero Bench Time refrigerant design
  • Expanded polystyrene insulation
  • Low packaging material cost
  • Simple designs
  • Air flow channels built into EPS, allows for less refrigerant needed in each packout
Duration
  • Refrigerated and CRT: Minimum of two days
  • Frozen: Minimum of four days
Shipping pharmaceuticals, clinical treatments and diagnostic samples
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 425 x 400 x 413 mm (16.7 x 15.7 x 16.2 in.)
Capacity (English) 0.66 gal.
Weight (English) 23.8 lb.
Weight (Metric) 10.8 kg
Material Expanded Polystyrene
Type Insulated Shipper
Quantity 24 Pallets
Capacity (Metric) 2.5 L
Temperature Range <-20°C