Description
- Reversed lozenge finish further enhances levels of grip.
- Longer length (38cm) than standard, extending protection to the wrist and lower forearm area
- Flocked lining and flexible nitrile file offer exceptional comfort
- Gauntlet style
- Reusable
- Latex free
- Resistant to chemicals and liquid protection
- CE, EN374-3:2003, CATEGORY III
Specifications
Specifications
| Back Material | Nitrile |
| Cuff Material | Nitrile |
| Applications | Industrial |
| Color | Green |
| Exterior Finish | Smooth |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 15 mil |
| Palm Thickness (Metric) | 0.38 mm |
| Product Type | Immersion Gloves |
| Show More |
Product Title
Haben Sie Verbesserungsvorschläge?