missing translation for 'onlineSavingsMsg'
Learn More

Solifenacin Succinate, TRC

Product Code. 30136053
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136053 50 mg 50mg
30133506 1 g 1g
17943025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136053 Supplier Toronto Research Chemicals Supplier No. TRCS67670150MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 242478-38-2
Molecular Formula C23 H26 N2 O2 . C4 H6 O4
Molecular Weight (g/mol) 480.55
Synonym Solifenacin succinate, (3R)-1-Azabicyclo[2.2.2]octan-3-yl (1S)-1-phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate hydrogen butanedioate, Butanedioic acid, compd. with (1S)-(3R)-1-azabicyclo[2.2.2]oct-3-yl 3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate (1:1), 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, (3R)-1-azabicyclo[2.2.2]oct-3-yl ester, (1S)-, butanedioate (1:1) (9CI), Vesicare, Vesikur, YM 67905
IUPAC Name [(3R)-1-azabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;butanedioic acid
SMILES OC(=O)CCC(=O)O.O=C(O[C@H]1CN2CCC1CC2)N3CCc4ccccc4[C@@H]3c5ccccc5

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H26N2O2.C4H6O4/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;5-3(6)1-2-4(7)8/h1-9,18,21-22H,10-16H2;1-2H2,(H,5,6)(H,7,8)/t21-,22-;/m0./s1
Formula Weight 480.226
Percent Purity >95%
Chemical Name or Material Solifenacin succinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.