Learn More
AMPri™ SolidSafety ChemN Special Nitrile Gloves
Product Code. 16637405
Change view
Click to view available options
Size:
Large
X-Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16637405 Large Pack of 100
16667405 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16637405 Supplier AMPri™ Supplier No. 81303L
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
This item is not returnable. Vea la política de devoluciones
SolidSafety ChemN Special Nitrile Protective Gloves are silicone-free and made of a special polymer that provides good chemical and physical resistance, as well as proven resistance to microorganisms.
  • Category III
  • Medical device Class I
  • Food grade
  • Chemical and physical resistance
  • Proven resistance to microorganisms
  • Silicone-free
  • Soft, comfortable fit, wearing comfort
  • Vera good grip, even if used in damp or wet environments
  • Non-sterile
  • Powder-free
  • Color: green
Especificaciones
Certifications/Compliance (Europe) EN 420, EN 455, EN 374/Type B (JKPT), Food grade according to Regulation (EU) 1935/2004, PPE Category III, Medical device Class I according to Regulation (EU) 2016/745
AQL 0.65
Color Green
Material Nitrile
Powdered Powder Free
Size Large
Length (Metric) 240 mm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.