Learn More
Hirschmann™ Solarus™ USB Connection Cable
Product Code. 10287312
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10287312 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10287312 Supplier Hirschmann™ Supplier No. 9564006
  • ONLINE EXCLUSIVE PRICE
    260.40 EUR
    164.48 EUR / Pack of 1
    Save 95.92 EUR
    36% Off
Estimated Shipment: 28-12-2026
For the USB interface between the unit and a PC.
  • Black in color
  • Length of 2 m
  • Accessory item
Specifications
Color Black
Length (Metric) 2 m
For Use With (Equipment) Solarus Titration Units
For Use With (Application) Data Transfer Between Titration Unit and PC
Type Connection Cable