Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Product Code. 16281581 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Dual Stack Universal Platform
Universal Platform
For Use With (Equipment):
Solaris 2000
Solaris 4000
Dimensions (L x W):
12 x 14 in.
18 x 18 in.
18 x 24 in.
18 x 30 in.
36 x 24 in.
Unit Size:
1 unit
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Dimensions (L x W) Unit Size
16281581 Universal Platform Solaris 2000 18 x 24 in. 1 unit
16221581 Universal Platform Solaris 2000 12 x 14 in. 1 unit
16231581 Dual Stack Universal Platform Solaris 2000 12 x 14 in. 1 unit
16251581 Universal Platform Solaris 2000 18 x 18 in. 1 unit
16261581 Dual Stack Universal Platform Solaris 2000 18 x 18 in. 1 unit
16201591 Dual Stack Universal Platform Solaris 4000 18 x 30 in. 1 unit
16291581 Universal Platform Solaris 4000 18 x 30 in. 1 unit
16231591 Universal Platform Solaris 4000 36 x 24 in. 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16281581 Supplier Thermo Scientific™ Supplier No. SK1824
  • ONLINE EXCLUSIVE PRICE
    585.00 EUR
    474.30 EUR / Pack of 1
    Save 110.70 EUR
    18% Off
View all promotions
Estimated Shipment: 04-05-2027
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab. Select a platform and clamp configuration that best meets your application, whether you require a single vessel type and size.
Certifications
UL, cUL, CE
Warranty
1 Year
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Platform
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W) 18 x 24 in.
Length (English) Platform 18 in.
Length (Metric) Platform 457.2 mm
Width (English) Platform 24 in.
Width (Metric) Platform 610 mm
Model Solaris Shakers
For Use With (Equipment) Solaris 2000
Includes Platform
Product Line Solaris
Warranty and Service Offering 1 Year