Learn More
Thermo Scientific™ SOLA™ SPE Plates
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
275.78 EUR - 555.00 EUR
Product Code | Brand | Description | Sorbent | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Sorbent | Price | Quantity & Availability | ||||
11899163
|
Thermo Scientific™
60309-001 |
10 mg/2 mL; SOLA 96-well SPE plate; 1 Pk. | HRP (Reversed Phase Polymeric) |
275.78 EUR
Each |
In Stock
Log in to see stock. |
||||
16697845
|
Thermo Scientific™
60509-001 |
SOLA HRP SPE 30 mg/2 mL 96-well plate | HRP (Reversed Phase Polymeric) |
318.59 EUR
1 pack |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
15547495
|
Thermo Scientific™
60309-002 |
10 mg/2 mL; SOLA-CX 96 well SPE plate; 1 Pk. | Mixed Mode Strong Cation Exchanger, Polymeric |
284.61 EUR
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16607855
|
Thermo Scientific™
60509-002 |
SOLA SCX SPE 30 mg/2 mL 96-well plate | Mixed Mode Strong Cation Exchanger, Polymeric |
328.61 EUR
1 pack |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
15550163
|
Thermo Scientific™
60309-003 |
10 mg/2 mL; SOLA-AX 96 well SPE plate; 1 Pk. | Mixed Mode Strong Anion Exchanger, Polymeric |
350.80 EUR
Each |
In Stock
Log in to see stock. |
||||
16617855
|
Thermo Scientific™
60509-003 |
SOLA SAX SPE 30 mg/2 mL 96-well plate | Mixed Mode Strong Anion Exchanger, Polymeric |
328.61 EUR
1 pack |
In Stock
Log in to see stock. |
||||
13493038
|
Thermo Scientific™
60309-004 |
10 mg/2 mL; SOLA WCX 96 well SPE plate; 1 Pk. | Mixed Mode Weak Cation Exchanger, Polymeric |
288.20 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
16627855
|
Thermo Scientific™
60509-004 |
SOLA WCX SPE 30 mg/2 mL 96-well plate | Mixed Mode Weak Cation Exchanger, Polymeric |
332.76 EUR
1 pack |
In Stock
Log in to see stock. |
||||
15179653
|
Thermo Scientific™
60309-005 |
10 mg/2 mL; SOLA WAX 96 well SPE plate; 1 Pk. | Mixed Mode Weak Anion Exchanger, Polymeric |
284.61 EUR
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
16637855
|
Thermo Scientific™
60509-005 |
SOLA WAX SPE 30 mg/2 mL 96-well plate | Mixed Mode Weak Anion Exchanger, Polymeric |
328.61 EUR
1 pack |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
11819173
|
Thermo Scientific™
60309-003 |
10 mg/2 mL; SOLA-AX 96 well SPE plate; 1 Pk. | Mixed Mode Strong Anion Exchanger, Polymeric |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11809173
|
Thermo Scientific™
60309-002 |
10 mg/2 mL; SOLA-CX 96 well SPE plate; 1 Pk. | Mixed Mode Strong Cation Exchanger, Polymeric |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
- Higher levels of reproducibility
- Lower elution volumes
- High extract cleanliness
- 10mg/2mL 96 well plate format
- Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
- SOLA products are also available in cartridge format.
Specifications
SOLA | |
96-well SPE Plate |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.